Assay Details
| Gene Symbol: POU1F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122757.2 | NP_001116229.1 | 1 - 2 | 263 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001585.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
cell fate specification
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
B cell differentiation
positive regulation of inositol trisphosphate biosynthetic process
positive regulation of multicellular organism growth
regulation of insulin-like growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nuclear transport
somatotropin secreting cell development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
RNA polymerase II transcription coactivator activity
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
negative regulation of transcription from RNA polymerase II promoter
cell fate specification
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
B cell differentiation
positive regulation of inositol trisphosphate biosynthetic process
positive regulation of multicellular organism growth
regulation of insulin-like growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nuclear transport
somatotropin secreting cell development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
RNA polymerase II transcription coactivator activity
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03028650.hg.1 | TC03001585.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028650.hg.1 | ENST00000344265.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028650.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028652.hg.1 | TC03001585.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028652.hg.1 | ENST00000344265.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028652.hg.1 | ENST00000350375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028652.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028652.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028653.hg.1 | TC03001585.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028653.hg.1 | ENST00000350375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028653.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028653.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028651.hg.1 | TC03001585.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028651.hg.1 | ENST00000344265.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028651.hg.1 | ENST00000350375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028651.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028651.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028651.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187185.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187185.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187185.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187185.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187185.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187185.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187185.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187185.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187185.hg.1 | TC0300011712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300187182.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187182.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187182.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187182.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187182.hg.1 | TC0300011712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187186.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187186.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187186.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187186.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187186.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187186.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187186.hg.1 | TC0300011712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | ENST00000344265.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | ENST00000350375.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | ENST00000560656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | ENST00000561167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | NM_000306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | NM_001122757.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187184.hg.1 | TC0300011712.hg.1 | Clariom D TC |

POU1F1
Genomic Map