Assay Details
| Gene Symbol: EPHA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288629.1 | NP_001275558.1 | 8 - 9 | 1966 | 66 | |
| NM_004440.3 | NP_004431.1 | 8 - 9 | 1981 | 66 | ||
| XM_005248669.2 | XP_005248726.1 | 8 - 9 | 2038 | 66 | ||
| XM_005248671.2 | XP_005248728.1 | 8 - 9 | 2038 | 66 | ||
| XM_017010365.1 | XP_016865854.1 | 8 - 9 | 2023 | 66 | ||
| GenBank mRNA | AB209269.1 | - | 8 - 9 | 1953 | 66 | |
| AK313529.1 | - | 8 - 9 | 1953 | 66 | ||
| BC126125.1 | - | 8 - 9 | 1855 | 8991841 | 66 | |
| BC126151.1 | - | 8 - 9 | 1855 | 8991867 | 66 | |
| BC143857.1 | - | 8 - 9 | 1840 | 9052373 | 66 | |
| BC143858.1 | - | 8 - 9 | 1855 | 9052374 | 66 | |
| L36642.1 | - | 8 - 9 | 1925 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001952.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
apoptotic process
brain development
phosphorylation
peptidyl-tyrosine phosphorylation
regulation of cell-cell adhesion
retinal ganglion cell axon guidance
regulation of protein autophosphorylation
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of neuron apoptotic process
ephrin receptor signaling pathway
negative regulation of collateral sprouting
branching morphogenesis of a nerve
regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
negative regulation of synapse assembly
regulation of ERK1 and ERK2 cascade
nephric duct morphogenesis
protein tyrosine kinase activity
GPI-linked ephrin receptor activity
protein binding
ATP binding
axon guidance receptor activity
chemorepellent activity
ephrin receptor binding
positive regulation of protein phosphorylation
apoptotic process
brain development
phosphorylation
peptidyl-tyrosine phosphorylation
regulation of cell-cell adhesion
retinal ganglion cell axon guidance
regulation of protein autophosphorylation
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of neuron apoptotic process
ephrin receptor signaling pathway
negative regulation of collateral sprouting
branching morphogenesis of a nerve
regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
negative regulation of synapse assembly
regulation of ERK1 and ERK2 cascade
nephric duct morphogenesis
protein tyrosine kinase activity
GPI-linked ephrin receptor activity
protein binding
ATP binding
axon guidance receptor activity
chemorepellent activity
ephrin receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024033.hg.1 | TC06001952.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024033.hg.1 | ENST00000369303.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024033.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_001288630.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012597.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186818.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186818.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186818.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186818.hg.1 | ENST00000369303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186818.hg.1 | NM_001288629.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186818.hg.1 | NM_004440.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186818.hg.1 | TC0600012597.hg.1 | Clariom D TC |

EPHA7
Genomic Map