Assay Details
| Gene Symbol: EPHA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005248671.2 | XP_005248728.1 | 9 - 10 | 2090 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPHA7| Gene Symbol: EPHA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005248671.2 | XP_005248728.1 | 9 - 10 | 2090 | 129 | |
Set Membership: |
|