Assay Details
| Gene Symbol: ABHD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272069.1 | NP_001258998.1 | 3 - 4 | 620 | 109 | |
| NM_018394.3 | NP_060864.1 | 3 - 4 | 620 | 109 | ||
| NR_073570.1 | 2 - 3 | 436 | 109 | |||
| XM_011512960.2 | XP_011511262.1 | 3 - 4 | 444 | 109 | ||
| GenBank mRNA | AK300018.1 | - | 2 - 3 | 302 | 109 | |
| AL713726.1 | - | 2 - 3 | 218 | 109 | ||
| BC014516.2 | - | 3 - 4 | 459 | 3928510 | 109 | |
| DC395725.1 | - | 2 - 3 | 440 | 109 | ||
| DR760607.1 | - | 3 - 4 | 445 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000566.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK300018.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000497293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000497293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | ENST00000497293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010280.hg.1 | TC03000566.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010280.hg.1 | ENST00000273359.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010280.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010280.hg.1 | ENST00000534857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010280.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010282.hg.1 | TC03000566.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010282.hg.1 | ENST00000273359.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010282.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010282.hg.1 | ENST00000534857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010282.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159330.hg.1 | TC0300008331.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300159331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159331.hg.1 | TC0300008331.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000493784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000497293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000497293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | ENST00000497293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008331.hg.1 | NR_073571.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | ENST00000273359.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | ENST00000491580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | ENST00000494817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | NM_001272069.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | NM_018394.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | NR_073570.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159329.hg.1 | TC0300008331.hg.1 | Clariom D TC |

ABHD10
Genomic Map