Assay Details
| Gene Symbol: QSOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181701.3 | NP_859052.3 | 4 - 5 | 626 | 59 | |
| GenBank mRNA | AJ318051.1 | - | 4 - 5 | 626 | 59 | |
| AL834369.1 | - | 1 - 2 | 117 | 59 | ||
| BC047604.2 | - | 5 - 6 | 508 | 59 | ||
| BQ269870.1 | - | 4 - 5 | 583 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001726.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09022060.hg.1 | TC09001726.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022060.hg.1 | ENST00000358701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022060.hg.1 | ENST00000389471.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022060.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022058.hg.1 | TC09001726.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022058.hg.1 | ENST00000358701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022058.hg.1 | ENST00000389471.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022058.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022061.hg.1 | TC09001726.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022061.hg.1 | ENST00000358701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022061.hg.1 | ENST00000389471.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022061.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022062.hg.1 | TC09001726.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022062.hg.1 | ENST00000358701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022062.hg.1 | ENST00000389471.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022062.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022057.hg.1 | TC09001726.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022057.hg.1 | ENST00000358701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022057.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181871.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181871.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181871.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181871.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181871.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181871.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181871.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181871.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181871.hg.1 | TC0900011970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181873.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181873.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181873.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181873.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181873.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181873.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181873.hg.1 | TC0900011970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181876.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181876.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181876.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181876.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181876.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181876.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181876.hg.1 | TC0900011970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181870.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181870.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181870.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181870.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181870.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181870.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181870.hg.1 | TC0900011970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181874.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181874.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181874.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181874.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181874.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181874.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181874.hg.1 | TC0900011970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181875.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181875.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181875.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181875.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181875.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181875.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181875.hg.1 | TC0900011970.hg.1 | Clariom D TC |

QSOX2
Genomic Map