Assay Details
| Gene Symbol: HNRNPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534546.1 | XP_011532848.1 | 2 - 3 | 423 | 75 | |
| XM_017009417.1 | XP_016864906.1 | 2 - 3 | 423 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011534546.1, XM_017009417.1 transcripts.
Set Membership: |
|

HNRNPH1
Genomic Map