Assay Details
| Gene Symbol: SMG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005256569.3 | XP_005256626.1 | 1 - 2 | 347 | 110 | |
| XM_011523769.2 | XP_011522071.1 | 2 - 3 | 337 | 110 | ||
| GenBank mRNA | AK024131.1 | - | 1 - 2 | 512 | 110 | |
| AU120859.1 | - | 1 - 2 | 512 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK024131.1, XM_005256569.3, XM_011523769.2 transcripts.
Set Membership: |
|

SMG6
Genomic Map