Assay Details
| Gene Symbol: HK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000189.4 | NP_000180.2 | 3 - 4 | 2304 | 69 | |
| XM_005264280.1 | XP_005264337.1 | 3 - 4 | 784 | 69 | ||
| XM_011532807.1 | XP_011531109.1 | 3 - 4 | 784 | 69 | ||
| XM_017003945.1 | XP_016859434.1 | 3 - 4 | 784 | 69 | ||
| GenBank mRNA | AF148513.1 | - | 3 - 4 | 827 | 69 | |
| AK290357.1 | - | 3 - 4 | 807 | 69 | ||
| BC021116.1 | - | 3 - 4 | 822 | 4652321 | 69 | |
| BC064369.1 | - | 3 - 4 | 820 | 5586773 | 69 | |
| CR749202.1 | - | 3 - 4 | 484 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000466.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular glucose homeostasis
response to ischemia
glycolytic process
lactation
apoptotic mitochondrial changes
glucose transport
negative regulation of mitochondrial membrane permeability
regulation of glucose import
carbohydrate phosphorylation
glucose 6-phosphate metabolic process
canonical glycolysis
establishment of protein localization to mitochondrion
maintenance of protein location in mitochondrion
positive regulation of mitophagy in response to mitochondrial depolarization
negative regulation of reactive oxygen species metabolic process
glucokinase activity
hexokinase activity
protein binding
ATP binding
glucose binding
fructokinase activity
mannokinase activity
cellular glucose homeostasis
response to ischemia
glycolytic process
lactation
apoptotic mitochondrial changes
glucose transport
negative regulation of mitochondrial membrane permeability
regulation of glucose import
carbohydrate phosphorylation
glucose 6-phosphate metabolic process
canonical glycolysis
establishment of protein localization to mitochondrion
maintenance of protein location in mitochondrion
positive regulation of mitophagy in response to mitochondrial depolarization
negative regulation of reactive oxygen species metabolic process
glucokinase activity
hexokinase activity
protein binding
ATP binding
glucose binding
fructokinase activity
mannokinase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02007347.hg.1 | TC02000466.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007347.hg.1 | ENST00000290573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007347.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007347.hg.1 | ENST00000535740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007347.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007345.hg.1 | TC02000466.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007345.hg.1 | ENST00000290573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007345.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007345.hg.1 | ENST00000535740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007345.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156303.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156303.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156303.hg.1 | ENST00000472302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156303.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156303.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156303.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156303.hg.1 | ENST00000472302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156303.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156303.hg.1 | TC0200008099.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | ENST00000472302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008099.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156305.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156305.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156305.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156305.hg.1 | ENST00000290573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156305.hg.1 | ENST00000409174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156305.hg.1 | NM_000189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156305.hg.1 | TC0200008099.hg.1 | Clariom D TC |

HK2
Genomic Map