Assay Details
| Gene Symbol: KIF1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006612.5 | NP_006603.2 | 11 - 12 | 1297 | 51 | |
| XM_005256424.2 | XP_005256481.1 | 12 - 13 | 1288 | 51 | ||
| GenBank mRNA | AB014606.2 | - | 11 - 12 | 1293 | 51 | |
| AK307401.1 | - | 11 - 12 | 1140 | 51 | ||
| BC034993.1 | - | 11 - 12 | 1271 | 5266798 | 51 | |
| U91329.1 | - | 10 - 11 | 1053 | 51 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000060.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000704.hg.1 | ENST00000320785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000704.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000704.hg.1 | TC17000060.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700145770.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145770.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700145770.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145770.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700145770.hg.1 | TC1700006660.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |

KIF1C
Genomic Map