Assay Details
| Gene Symbol: ACTR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005735.3 | NP_005726.1 | 1 - 2 | 255 | 75 | |
| XM_005263854.4 | XP_005263911.1 | 1 - 2 | 90 | 75 | ||
| GenBank mRNA | BC004374.2 | - | 1 - 2 | 123 | 3638252 | 75 |
| BC006372.2 | - | 1 - 2 | 88 | 4138683 | 75 | |
| BC010090.2 | - | 1 - 2 | 155 | 2989545 | 75 | |
| X82207.1 | - | 1 - 2 | 103 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002121.hg
Additional Information:
This assay is in the 5'UTR region of the XM_005263854.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02034475.hg.1 | TC02002121.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034475.hg.1 | ENST00000289228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034475.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034474.hg.1 | TC02002121.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034474.hg.1 | ENST00000289228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034474.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034470.hg.1 | TC02002121.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034470.hg.1 | ENST00000289228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034470.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000460427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000460427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000460427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000460427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | ENST00000460427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034471.hg.1 | TC02002121.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034471.hg.1 | ENST00000289228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034471.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034473.hg.1 | TC02002121.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034473.hg.1 | ENST00000289228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034473.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199458.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199458.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199458.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199458.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199458.hg.1 | TC0200013573.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200199455.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199455.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199455.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199455.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199455.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199455.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199455.hg.1 | TC0200013573.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200199457.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199457.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199457.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199457.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199457.hg.1 | TC0200013573.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200199453.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199453.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199453.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199453.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199453.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199453.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199453.hg.1 | TC0200013573.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200199454.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199454.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199454.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199454.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199454.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199454.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199454.hg.1 | TC0200013573.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200199459.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199459.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199459.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200199459.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200199459.hg.1 | TC0200013573.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000289228.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000451664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000460427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000460427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000460427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000460427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | ENST00000460427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013573.hg.1 | NM_005735.3 | Refseq |

ACTR1B
Genomic Map