Assay Details
| Gene Symbol: ODF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014440.3 | NP_001014440.2 | 4 - 5 | 551 | 55 | |
| GenBank mRNA | BC127935.1 | - | 3 - 4 | 380 | 55 | |
| BG149596.1 | - | 2 - 3 | 284 | 55 | ||
| BM973820.1 | - | 3 - 4 | 55 | |||
| CD674272.1 | - | 3 - 4 | 450 | 55 | ||
| S72487.1 | - | 1 - 2 | 624 | 55 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001501.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22015675.hg.1 | TC22001501.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015675.hg.1 | ENST00000329363.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015675.hg.1 | ENST00000401779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015675.hg.1 | ENST00000403326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015675.hg.1 | ENST00000405135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015675.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015675.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | TC2200009363.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170813.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |

ODF3B
Genomic Map