Assay Details
| Gene Symbol: CATX-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF083117.1 | - | 1 | 62 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC285000 | LOC285000 | -272.4kb | Sense | Chr. 2 - 105593097 - 105610559 on NCBI Build GRCh38 | |
| Downstream | chromosome 2 open reading frame 40 | C2orf40 | ECRG4 | 179.9kb | Anti-sense | Chr. 2 - 106063290 - 106078155 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CATX-1
Genomic Map