Assay Details
| Gene Symbol: CCNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033670.2 | NP_391990.1 | 3 - 4 | 319 | 92 | |
| GenBank mRNA | AJ314765.1 | - | 3 - 4 | 323 | 92 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000491907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000491907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000491907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000491907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000491907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000493507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000493507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000493507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000493507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | ENST00000493507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000276014.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000348603.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000376042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000476167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000487204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000491907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000491907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000491907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000491907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000491907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000493507.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000493507.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000493507.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000493507.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | ENST00000493507.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033031.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007251.hg.1 | NM_033670.2 | Refseq |

CCNB3
Genomic Map