Assay Details
| Gene Symbol: SRRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110199.2 | NP_001103669.1 | 3 - 4 | 556 | 125 | |
| NM_001291831.1 | NP_001278760.1 | 3 - 4 | 556 | 125 | ||
| GenBank mRNA | BC029292.1 | - | 3 - 4 | 549 | 125 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000483.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07006736.hg.1 | TC07000483.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006736.hg.1 | ENST00000326382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006736.hg.1 | ENST00000388802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006736.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156061.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156061.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156061.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156061.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156061.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156061.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156061.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156061.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156061.hg.1 | TC0700008088.hg.1 | Clariom D TC |

SRRM3
Genomic Map