| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000372037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000480152.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000480152.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000480152.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000480152.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
ENST00000480152.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008315.hg.1 |
NM_004329.2 |
Refseq |