Assay Details
| Gene Symbol: ATG4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321289.1 | NP_001308218.1 | 9 - 10 | 1094 | 66 | |
| NM_178270.3 | NP_840054.1 | 8 - 9 | 792 | 66 | ||
| XM_011530841.2 | XP_011529143.1 | 8 - 9 | 693 | 66 | ||
| GenBank mRNA | AJ320509.1 | - | 8 - 9 | 650 | 66 | |
More Information
Set Membership: |
|

ATG4A
Genomic Map