Assay Details
| Gene Symbol: ITGAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002209.2 | NP_002200.2 | 6 - 7 | 745 | 123 | |
| XM_005255313.1 | XP_005255370.1 | 6 - 7 | 665 | 123 | ||
| XM_011545849.1 | XP_011544151.1 | 6 - 7 | 678 | 123 | ||
| GenBank mRNA | AK314246.1 | - | 6 - 7 | 699 | 123 | |
| Y00796.1 | - | 6 - 7 | 657 | 123 | ||
More Information
Set Membership: |
|

ITGAL
Genomic Map