Assay Details
| Gene Symbol: RASIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017805.2 | NP_060275.2 | 1 - 2 | 196 | 99 | |
| XM_011527053.2 | XP_011525355.2 | 1 - 2 | 316 | 99 | ||
| XM_011527054.2 | XP_011525356.2 | 1 - 2 | 316 | 99 | ||
| XM_011527055.2 | XP_011525357.2 | 1 - 2 | 316 | 99 | ||
| XM_017026914.1 | XP_016882403.1 | 1 - 2 | 316 | 99 | ||
| XM_017026915.1 | XP_016882404.1 | 1 - 2 | 316 | 99 | ||
| GenBank mRNA | BC042111.1 | - | 1 - 2 | 204 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001693.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC042111.1,NM_017805.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023098.hg.1 | ENST00000222145.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023098.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023098.hg.1 | TC19001693.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023097.hg.1 | ENST00000222145.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023097.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023097.hg.1 | TC19001693.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023099.hg.1 | ENST00000222145.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023099.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023099.hg.1 | TC19001693.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900192769.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192769.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192769.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192769.hg.1 | TC1900011094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192769.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192769.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192769.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192767.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192767.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192767.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192767.hg.1 | TC1900011094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192767.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192767.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192767.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192771.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192771.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192771.hg.1 | TC1900011094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192771.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192771.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |

RASIP1
Genomic Map