Assay Details
| Gene Symbol: RASIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017805.2 | NP_060275.2 | 2 - 3 | 327 | 98 | |
| XM_011527053.2 | XP_011525355.2 | 2 - 3 | 447 | 98 | ||
| XM_011527054.2 | XP_011525356.2 | 2 - 3 | 447 | 98 | ||
| XM_011527055.2 | XP_011525357.2 | 2 - 3 | 447 | 98 | ||
| XM_017026914.1 | XP_016882403.1 | 2 - 3 | 447 | 98 | ||
| XM_017026915.1 | XP_016882404.1 | 2 - 3 | 447 | 98 | ||
| GenBank mRNA | AY378097.1 | - | 2 - 3 | 195 | 98 | |
| BC028614.1 | - | 2 - 3 | 199 | 4815880 | 98 | |
| BC042111.1 | - | 2 - 3 | 335 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001693.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19023095.hg.1 | ENST00000222145.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023095.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023095.hg.1 | TC19001693.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023096.hg.1 | ENST00000222145.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023096.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023096.hg.1 | TC19001693.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | TC1900011094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192763.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192765.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192765.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192765.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192765.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192765.hg.1 | TC1900011094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192765.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192765.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192765.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192765.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192766.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192766.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192766.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192766.hg.1 | TC1900011094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192766.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192766.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192766.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |

RASIP1
Genomic Map