Assay Details
| Gene Symbol: RASIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527053.2 | XP_011525355.2 | 11 - 12 | 3007 | 138 | |
| XM_011527055.2 | XP_011525357.2 | 8 - 9 | 2713 | 138 | ||
More Information
Set Membership: |
|

RASIP1
Genomic Map