Assay Details
| Gene Symbol: RASIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017805.2 | NP_060275.2 | 5 - 6 | 2026 | 60 | |
| XM_011527053.2 | XP_011525355.2 | 5 - 6 | 2146 | 60 | ||
| XM_011527054.2 | XP_011525356.2 | 5 - 6 | 2146 | 60 | ||
| XM_017026914.1 | XP_016882403.1 | 5 - 6 | 2146 | 60 | ||
| GenBank mRNA | AK000408.1 | - | 1 - 2 | 141 | 60 | |
| AY378097.1 | - | 5 - 6 | 1891 | 60 | ||
| BC021860.1 | - | 1 - 2 | 156 | 60 | ||
| BC028614.1 | - | 5 - 6 | 1898 | 4815880 | 60 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001693.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK000408.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19023093.hg.1 | ENST00000222145.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023093.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023093.hg.1 | TC19001693.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023094.hg.1 | ENST00000222145.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023094.hg.1 | TC19001693.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900192760.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192760.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192760.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192760.hg.1 | TC1900011094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192760.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192760.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192760.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192761.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192761.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192761.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192761.hg.1 | TC1900011094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192761.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192761.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192761.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192762.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192762.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192762.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192762.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192762.hg.1 | TC1900011094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192762.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192762.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192762.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192762.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000222145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000594232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000599291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000601530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | ENST00000621604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011094.hg.1 | NM_017805.2 | Refseq |

RASIP1
Genomic Map