Assay Details
| Gene Symbol: RASIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017805.2 | NP_060275.2 | 6 - 7 | 2063 | 107 | |
| XM_011527053.2 | XP_011525355.2 | 6 - 7 | 2183 | 107 | ||
| XM_011527054.2 | XP_011525356.2 | 6 - 7 | 2183 | 107 | ||
| XM_017026914.1 | XP_016882403.1 | 6 - 7 | 2183 | 107 | ||
| GenBank mRNA | AK000408.1 | - | 2 - 3 | 178 | 107 | |
| AY378097.1 | - | 6 - 7 | 1928 | 107 | ||
| BC021860.1 | - | 2 - 3 | 193 | 107 | ||
| BC028614.1 | - | 6 - 7 | 1935 | 4815880 | 107 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001693.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK000408.1 transcripts.
Set Membership: |
|

RASIP1
Genomic Map