Assay Details
| Gene Symbol: ZNF426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318055.1 | NP_001304984.1 | 3 - 4 | 311 | 104 | |
| NM_001318056.1 | NP_001304985.1 | 3 - 4 | 311 | 104 | ||
| NM_024106.2 | NP_077011.1 | 3 - 4 | 311 | 104 | ||
| GenBank mRNA | AK095759.1 | - | 1 - 2 | 366 | 104 | |
| BC001791.1 | - | 3 - 4 | 293 | 3543910 | 104 | |
| BE315219.1 | - | 3 - 4 | 256 | 104 | ||
| BP245135.1 | - | 3 - 4 | 311 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001148.hg
Additional Information:
This assay is in the 5'UTR region of the AK095759.1,BC001791.1,NM_001318055.1,NM_001318056.1,NM_024106.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19015823.hg.1 | ENST00000253115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015823.hg.1 | ENST00000535489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015823.hg.1 | ENST00000545189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015823.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015823.hg.1 | TC19001148.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015824.hg.1 | ENST00000253115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015824.hg.1 | ENST00000535489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015824.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015824.hg.1 | TC19001148.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900177176.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177176.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177176.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177176.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900177176.hg.1 | TC1900009586.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900177176.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177176.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177176.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177176.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009586.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | NM_024106.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | TC1900009586.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | ENST00000253115.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | ENST00000535489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | ENST00000589289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | ENST00000593003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | NM_001300883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900177175.hg.1 | NM_024106.2 | Refseq |

ZNF426
Genomic Map