Assay Details
| Gene Symbol: CCDC47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020198.2 | NP_064583.2 | 8 - 9 | 1287 | 127 | |
| XM_005257527.2 | XP_005257584.1 | 8 - 9 | 1155 | 127 | ||
| GenBank mRNA | AF113221.1 | - | 6 - 7 | 634 | 127 | |
| AF226054.1 | - | 8 - 9 | 1135 | 127 | ||
| AK027786.1 | - | 8 - 9 | 1129 | 127 | ||
| AK027844.1 | - | 8 - 9 | 1116 | 127 | ||
| AK075387.1 | - | 9 - 10 | 1216 | 127 | ||
| AK225813.1 | - | 8 - 9 | 1155 | 127 | ||
| AK314328.1 | - | 8 - 9 | 1142 | 127 | ||
| BC008905.2 | - | 8 - 9 | 1109 | 3688068 | 127 | |
| BC013600.2 | - | 5 - 6 | 578 | 127 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001785.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17023775.hg.1 | ENST00000225726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023775.hg.1 | ENST00000403162.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023775.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023775.hg.1 | TC17001785.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000580986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000580986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000580986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000584112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000584112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000584112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000584112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700207124.hg.1 | TC1700012446.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000225726.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000403162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000580986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000580986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000580986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000582331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000584112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000584112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000584112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000584112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | ENST00000612558.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012446.hg.1 | NM_020198.2 | Refseq |

CCDC47
Genomic Map