Assay Details
| Gene Symbol: RPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001021.4 | NP_001012.1 | 1 - 2 | 123 | 73 | |
| NR_111944.1 | 1 - 2 | 123 | 73 | |||
| GenBank mRNA | BC062715.1 | - | 1 - 2 | 121 | 4068842 | 73 |
| BM017772.1 | - | 1 - 2 | 124 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062715.1, NM_001021.4 transcripts.
Set Membership: |
|

RPS17
Genomic Map