Assay Details
| Gene Symbol: ETS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256295.1 | NP_001243224.1 | 7 - 8 | 1052 | 117 | |
| NM_005239.5 | NP_005230.1 | 6 - 7 | 882 | 117 | ||
| XM_005260935.1 | XP_005260992.1 | 6 - 7 | 955 | 117 | ||
| XM_017028290.1 | XP_016883779.1 | 6 - 7 | 3552 | 117 | ||
| GenBank mRNA | AK096841.1 | - | 7 - 8 | 1052 | 117 | |
| AK315563.1 | - | 6 - 7 | 785 | 117 | ||
| BC017040.1 | - | 6 - 7 | 754 | 3852274 | 117 | |
| BC042954.2 | - | 6 - 7 | 784 | 5276704 | 117 | |
| BT006838.1 | - | 5 - 6 | 592 | 117 | ||
| J04102.1 | - | 6 - 7 | 883 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000169.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
ectodermal cell fate commitment
transcription, DNA-templated
mesoderm development
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
primitive streak formation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
glucocorticoid receptor binding
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
ectodermal cell fate commitment
transcription, DNA-templated
mesoderm development
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
primitive streak formation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
glucocorticoid receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21001617.hg.1 | ENST00000360214.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001617.hg.1 | ENST00000360938.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001617.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001617.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001617.hg.1 | TC21000169.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001618.hg.1 | ENST00000360214.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001618.hg.1 | ENST00000360938.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001618.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001618.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001618.hg.1 | TC21000169.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | TC2100007140.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148163.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148164.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148164.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148164.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148164.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148164.hg.1 | TC2100007140.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100148164.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148164.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148164.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148164.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360214.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000360938.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000432278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | ENST00000456966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_001256295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007140.hg.1 | NM_005239.5 | Refseq |

ETS2
Genomic Map