Assay Details
| Gene Symbol: PTTG1IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004339.3 | NP_004330.1 | 4 - 5 | 737 | 134 | |
| NR_104597.1 | 3 - 4 | 628 | 134 | |||
| GenBank mRNA | AF149785.1 | - | 4 - 5 | 675 | 134 | |
| AK094882.1 | - | 5 - 6 | 442 | 134 | ||
| AK290139.1 | - | 4 - 5 | 490 | 131 | ||
| AK299204.1 | - | 2 - 3 | 296 | 134 | ||
| BC000415.2 | - | 4 - 5 | 494 | 2961420 | 134 | |
| BC012858.1 | - | 4 - 5 | 520 | 3857505 | 134 | |
| BC019295.1 | - | 4 - 5 | 484 | 2823814 | 134 | |
| BC020983.1 | - | 4 - 5 | 501 | 3851447 | 134 | |
| BC031097.1 | - | 4 - 5 | 515 | 5288388 | 134 | |
| BC034250.1 | - | 4 - 5 | 509 | 3862830 | 134 | |
| CN295711.1 | - | 3 - 4 | 419 | 134 | ||
| L48984.1 | - | 1 - 2 | 135 | 134 | ||
| Z50022.1 | - | 4 - 5 | 543 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

PTTG1IP
Genomic Map