Assay Details
| Gene Symbol: TNNI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000363.4 | NP_000354.4 | 6 - 7 | 514 | 77 | |
| GenBank mRNA | AK223451.1 | - | 6 - 7 | 514 | 77 | |
| BC096165.1 | - | 6 - 7 | 440 | 40003354 | 77 | |
| BC096166.1 | - | 6 - 7 | 440 | 40003355 | 77 | |
| BC096167.3 | - | 6 - 7 | 440 | 40003357 | 77 | |
| BC099631.3 | - | 6 - 7 | 440 | 40003359 | 77 | |
| BT019517.1 | - | 4 - 5 | 371 | 77 | ||
| CR541985.1 | - | 4 - 5 | 371 | 77 | ||
| CR542012.1 | - | 4 - 5 | 371 | 77 | ||
| EU747720.1 | - | 6 - 7 | 514 | 77 | ||
| M64247.1 | - | 6 - 7 | 379 | 77 | ||
| X54163.1 | - | 6 - 7 | 460 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001859.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vasculogenesis
regulation of systemic arterial blood pressure by ischemic conditions
skeletal muscle contraction
cellular calcium ion homeostasis
regulation of smooth muscle contraction
heart development
muscle filament sliding
negative regulation of ATPase activity
ventricular cardiac muscle tissue morphogenesis
heart contraction
cardiac muscle contraction
regulation of cardiac conduction
actin binding
protein binding
calcium channel inhibitor activity
protein kinase binding
protein domain specific binding
troponin C binding
troponin T binding
metal ion binding
calcium-dependent protein binding
vasculogenesis
regulation of systemic arterial blood pressure by ischemic conditions
skeletal muscle contraction
cellular calcium ion homeostasis
regulation of smooth muscle contraction
heart development
muscle filament sliding
negative regulation of ATPase activity
ventricular cardiac muscle tissue morphogenesis
heart contraction
cardiac muscle contraction
regulation of cardiac conduction
actin binding
protein binding
calcium channel inhibitor activity
protein kinase binding
protein domain specific binding
troponin C binding
troponin T binding
metal ion binding
calcium-dependent protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000586858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025276.hg.1 | ENST00000344887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025276.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025276.hg.1 | TC19001859.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025277.hg.1 | ENST00000344887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025277.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025277.hg.1 | TC19001859.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | TC1900012047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208189.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | TC1900012047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208194.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000344887.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000585806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000586858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000587176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000588882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000589864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | ENST00000590463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012047.hg.1 | NM_000363.4 | Refseq |

TNNI3
Genomic Map