Assay Details
| Gene Symbol: NUPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005249592.1 | XP_005249649.1 | 6 - 7 | 804 | 87 | |
| XM_005249593.3 | XP_005249650.1 | 6 - 7 | 1484 | 87 | ||
| XM_011515104.2 | XP_011513406.1 | 5 - 6 | 972 | 87 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
mRNA export from nucleus
tRNA export from nucleus
protein export from nucleus
mitotic nuclear envelope disassembly
regulation of glucose transport
viral process
protein sumoylation
viral transcription
gene silencing by RNA
intracellular transport of virus
regulation of cellular response to heat
nuclear export signal receptor activity
protein binding
poly(A) RNA binding
metal ion binding
mRNA export from nucleus
tRNA export from nucleus
protein export from nucleus
mitotic nuclear envelope disassembly
regulation of glucose transport
viral process
protein sumoylation
viral transcription
gene silencing by RNA
intracellular transport of virus
regulation of cellular response to heat
nuclear export signal receptor activity
protein binding
poly(A) RNA binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000410002.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000410002.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000410002.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000410002.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000486136.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000486136.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000486136.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000486136.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000486136.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000487595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000487595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000487595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000487595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000487595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000489145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000489145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000489145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000489145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000489145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000497500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000497500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000497500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000497500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000548367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000548367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | ENST00000548367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000258742.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000410002.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000410002.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000410002.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000410002.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000413919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000437140.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000438012.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000477844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000485250.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000486136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000486136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000486136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000486136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000486136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000487595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000487595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000487595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000487595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000487595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000489145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000489145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000489145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000489145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000489145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000497500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000497500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000497500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000497500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000548367.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000548367.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | ENST00000548367.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006910.hg.1 | NM_007342.2 | Refseq |

NUPL2
Genomic Map