Assay Details
| Gene Symbol: CSF2RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000395.2 | NP_000386.1 | 6 - 7 | 935 | 84 | |
| XM_005261340.3 | XP_005261397.1 | 6 - 7 | 994 | 84 | ||
| XM_011529903.2 | XP_011528205.1 | 6 - 7 | 994 | 84 | ||
| XM_011529904.2 | XP_011528206.1 | 6 - 7 | 994 | 84 | ||
| XM_011529905.2 | XP_011528207.1 | 6 - 7 | 994 | 84 | ||
| GenBank mRNA | AK302993.1 | - | 4 - 5 | 579 | 84 | |
| BC070085.1 | - | 6 - 7 | 935 | 30344148 | 84 | |
| CR456428.1 | - | 5 - 6 | 791 | 84 | ||
| M59941.1 | - | 5 - 6 | 746 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000271.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
signal transduction
respiratory gaseous exchange
peptidyl-tyrosine phosphorylation
cellular response to interleukin-3
interleukin-5-mediated signaling pathway
interleukin-3-mediated signaling pathway
positive regulation of GTPase activity
cellular protein metabolic process
protein tyrosine kinase activity
receptor activity
interleukin-3 receptor activity
interleukin-5 receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
MAPK cascade
signal transduction
respiratory gaseous exchange
peptidyl-tyrosine phosphorylation
cellular response to interleukin-3
interleukin-5-mediated signaling pathway
interleukin-3-mediated signaling pathway
positive regulation of GTPase activity
cellular protein metabolic process
protein tyrosine kinase activity
receptor activity
interleukin-3 receptor activity
interleukin-5 receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004989.hg.1 | ENST00000262825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004989.hg.1 | ENST00000403662.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004989.hg.1 | ENST00000406230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004989.hg.1 | ENST00000536485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004989.hg.1 | ENST00000539104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004989.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22004989.hg.1 | TC22000271.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | TC2200007273.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150944.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |

CSF2RB
Genomic Map