Assay Details
| Gene Symbol: CSF2RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005261340.3 | XP_005261397.1 | 7 - 8 | 1129 | 141 | |
| XM_011529903.2 | XP_011528205.1 | 7 - 8 | 1129 | 141 | ||
| XM_011529905.2 | XP_011528207.1 | 7 - 8 | 1129 | 141 | ||
| GenBank mRNA | AK302993.1 | - | 5 - 6 | 714 | 141 | |
| CR456428.1 | - | 6 - 7 | 926 | 141 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
signal transduction
respiratory gaseous exchange
peptidyl-tyrosine phosphorylation
cellular response to interleukin-3
interleukin-5-mediated signaling pathway
interleukin-3-mediated signaling pathway
positive regulation of GTPase activity
cellular protein metabolic process
protein tyrosine kinase activity
receptor activity
interleukin-3 receptor activity
interleukin-5 receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
MAPK cascade
signal transduction
respiratory gaseous exchange
peptidyl-tyrosine phosphorylation
cellular response to interleukin-3
interleukin-5-mediated signaling pathway
interleukin-3-mediated signaling pathway
positive regulation of GTPase activity
cellular protein metabolic process
protein tyrosine kinase activity
receptor activity
interleukin-3 receptor activity
interleukin-5 receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000262825.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000403662.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000406230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | ENST00000421539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007273.hg.1 | NM_000395.2 | Refseq |

CSF2RB
Genomic Map