Assay Details
| Gene Symbol: PRKAR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002736.2 | NP_002727.2 | 1 - 2 | 480 | 108 | |
| XM_011516398.1 | XP_011514700.1 | 1 - 2 | 485 | 108 | ||
| GenBank mRNA | AK291441.1 | - | 1 - 2 | 331 | 108 | |
| BC075800.1 | - | 1 - 2 | 519 | 30331962 | 108 | |
| CF593944.1 | - | 285 | 108 | |||
| M31158.1 | - | 1 - 2 | 471 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000688.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
renal water homeostasis
fatty acid metabolic process
blood coagulation
learning
activation of protein kinase A activity
intracellular signal transduction
cellular response to glucagon stimulus
response to antipsychotic drug
response to clozapine
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
protein domain specific binding
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
G2/M transition of mitotic cell cycle
renal water homeostasis
fatty acid metabolic process
blood coagulation
learning
activation of protein kinase A activity
intracellular signal transduction
cellular response to glucagon stimulus
response to antipsychotic drug
response to clozapine
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
protein domain specific binding
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07010728.hg.1 | TC07000688.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010728.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010728.hg.1 | ENST00000539794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010728.hg.1 | ENST00000543645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010728.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010730.hg.1 | TC07000688.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010730.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010730.hg.1 | ENST00000393613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010730.hg.1 | ENST00000539794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010730.hg.1 | ENST00000543645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010730.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010726.hg.1 | TC07000688.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010726.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010726.hg.1 | ENST00000539794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010726.hg.1 | ENST00000543645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010726.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010727.hg.1 | TC07000688.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010727.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010727.hg.1 | ENST00000543645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010727.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000488792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000488792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000488792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000488792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | ENST00000488792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010724.hg.1 | TC07000688.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010724.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07010724.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161731.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161731.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161731.hg.1 | TC0700008745.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161735.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161735.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161735.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161735.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161735.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161735.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161735.hg.1 | TC0700008745.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161733.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161733.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161733.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161733.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161733.hg.1 | TC0700008745.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700161732.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161732.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161732.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700161732.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700161732.hg.1 | TC0700008745.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000265717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000393613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000488792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000488792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000488792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000488792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | ENST00000488792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008745.hg.1 | NM_002736.2 | Refseq |

PRKAR2B
Genomic Map