Assay Details
| Gene Symbol: SND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014390.2 | NP_055205.2 | 16 - 17 | 2009 | 74 | |
| GenBank mRNA | AB209510.1 | - | 16 - 17 | 1947 | 74 | |
| AK096583.1 | - | 17 - 18 | 1934 | 74 | ||
| AK304181.1 | - | 16 - 17 | 1901 | 74 | ||
| AK309014.1 | - | 8 - 9 | 902 | 74 | ||
| AK312313.1 | - | 16 - 17 | 1963 | 74 | ||
| BC017180.2 | - | 16 - 17 | 1965 | 3345037 | 74 | |
| BE799345.1 | - | 3 - 4 | 345 | 74 | ||
| BT009785.1 | - | 16 - 17 | 1783 | 74 | ||
| U22055.1 | - | 16 - 17 | 1975 | 74 | ||
More Information
Set Membership: |
|

SND1
Genomic Map