Assay Details
| Gene Symbol: LSM14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011528607.2 | XP_011526909.1 | 6 - 7 | 1105 | 58 | |
| XM_011528611.2 | XP_011526913.1 | 4 - 5 | 793 | 58 | ||
| GenBank mRNA | BX370449.2 | - | 1 - 2 | 75 | 58 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |

LSM14B
Genomic Map