Assay Details
| Gene Symbol: HMGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB208998.1 | - | 4 | 1528 | 103 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010787.hg.1 | ENST00000429856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010787.hg.1 | ENST00000429856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000468384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000468384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000468384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000468384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000468384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000490788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000490788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | ENST00000490788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010787.hg.1 | ENST00000429856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010787.hg.1 | ENST00000429856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000326004.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000339872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000341423.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000399494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000405805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000468384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000468384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000468384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000468384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000468384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000490788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000490788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | ENST00000490788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008517.hg.1 | NM_002128.5 | Refseq |

HMGB1
Genomic Map