Assay Details
Gene Symbol: EHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB058779.2 | - | 22 | 5568 | 103 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700010424.hg.1 | ENST00000578020.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010424.hg.1 | ENST00000578020.1 | Ensembl |
