Assay Details
| Gene Symbol: CHST15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006717891.3 | XP_006717954.1 | 1 - 2 | 534 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006717891.3 transcripts.
Set Membership: |
|

CHST15
Genomic Map