Assay Details
| Gene Symbol: HK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322364.1 | NP_001309293.1 | 3 - 4 | 250 | 84 | |
| NM_033497.2 | NP_277032.1 | 4 - 5 | 372 | 84 | ||
| NM_033498.2 | NP_277033.1 | 4 - 5 | 426 | 84 | ||
| GenBank mRNA | DB088955.1 | - | 4 - 5 | 376 | 84 | |
More Information
Set Membership: |
|

HK1
Genomic Map