Assay Details
| Gene Symbol: SERPINA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085.4 | NP_001076.2 | 1 - 2 | 71 | 108 | |
| GenBank mRNA | AB209060.1 | - | 1 - 2 | 43 | 108 | |
| AK093049.1 | - | 4 - 5 | 955 | 108 | ||
| BC003559.1 | - | 1 - 2 | 44 | 3547133 | 108 | |
| BC010530.1 | - | 1 - 2 | 41 | 4152174 | 108 | |
| BC034554.1 | - | 1 - 2 | 35 | 4152390 | 108 | |
| BC070265.1 | - | 1 - 2 | 54 | 108 | ||
| BG565041.1 | - | 1 - 2 | 60 | 108 | ||
| CB111903.1 | - | 1 - 2 | 71 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14002309.hg
Additional Information:
This assay is in the 5'UTR region of the BC003559.1,BC010530.1,BC034554.1,BC070265.1,NM_001085.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14007307.hg.1 | TC14002309.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007307.hg.1 | ENST00000393078.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007307.hg.1 | ENST00000393080.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007307.hg.1 | ENST00000467132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007307.hg.1 | ENST00000468897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007307.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000485588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000485588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000556388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000556388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000556388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000621603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | ENST00000621603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007308.hg.1 | TC14002309.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007308.hg.1 | ENST00000393078.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007308.hg.1 | ENST00000393080.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007308.hg.1 | ENST00000467132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007308.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007301.hg.1 | TC14002309.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007301.hg.1 | ENST00000393078.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007301.hg.1 | ENST00000393080.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007301.hg.1 | ENST00000467132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007301.hg.1 | ENST00000468897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007301.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007306.hg.1 | TC14002309.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007306.hg.1 | ENST00000380354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007306.hg.1 | ENST00000393078.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007306.hg.1 | ENST00000393080.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007306.hg.1 | ENST00000467132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007306.hg.1 | ENST00000468897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007306.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180396.hg.1 | TC1400010644.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180395.hg.1 | TC1400010644.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000482740.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000485588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000485588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000556388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000556388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000556388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000621603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | ENST00000621603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010644.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000485588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000556388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000393078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000393080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000467132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000485588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000555820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000556388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | ENST00000556968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | NM_001085.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180388.hg.1 | TC1400010644.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 165806 | - | - |

SERPINA3
Genomic Map