Assay Details
| Gene Symbol: RARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318785.1 | NP_001305714.1 | 1 - 2 | 338 | 114 | |
| XM_017011072.1 | XP_016866561.1 | 1 - 2 | 283 | 114 | ||
| XM_017011075.1 | XP_016866564.1 | 1 - 2 | 283 | 114 | ||
| XM_017011076.1 | XP_016866565.1 | 1 - 2 | 283 | 114 | ||
| XM_017011079.1 | XP_016866568.1 | 1 - 2 | 283 | 114 | ||
| XM_017011080.1 | XP_016866569.1 | 1 - 2 | 283 | 114 | ||
| GenBank mRNA | AF143874.1 | - | 272 | 114 | ||
| BC105784.1 | - | 1 - 2 | 277 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001318785.1, XM_017011072.1, XM_017011075.1, XM_017011076.1, XM_017011079.1, XM_017011080.1 transcripts.
Set Membership: |
|

RARS2
Genomic Map