Assay Details
| Gene Symbol: DYNC1H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001376.4 | NP_001367.2 | 15 - 16 | 3733 | 114 | |
| GenBank mRNA | AB002323.3 | - | 15 - 16 | 3605 | 114 | |
| AB290157.1 | - | 15 - 16 | 3733 | 114 | ||
| U53530.1 | - | 2 - 3 | 183 | 114 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000777.hg.1
Set Membership: |
|

DYNC1H1
Genomic Map