Assay Details
| Gene Symbol: DYNC1H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001376.4 | NP_001367.2 | 57 - 58 | 11071 | 66 | |
| GenBank mRNA | AB002323.3 | - | 57 - 58 | 10943 | 66 | |
| AB290157.1 | - | 57 - 58 | 11071 | 66 | ||
| AL833600.1 | - | 29 - 30 | 5069 | 66 | ||
| BX648670.1 | - | 22 - 23 | 3673 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000777.hg.1
Set Membership: |
|

DYNC1H1
Genomic Map