Assay Details
| Gene Symbol: LINC01144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024270.1 | 1 | 1110 | 98 | ||
| GenBank mRNA | BC035497.2 | - | 1 | 1110 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger SWIM-type containing 5 | ZSWIM5 | -97.3kb | Anti-sense | Chr. 1 - 45016399 - 45206586 on NCBI Build GRCh38 | |
| Downstream | 4-hydroxyphenylpyruvate dioxygenase like | HPDL |
4-HPPD-L, GLOXD1 |
21.3kb | Sense | Chr. 1 - 45326873 - 45328675 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174832 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174809 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174822 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200088 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15055929 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200057 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174820 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174802 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200069 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174826 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174821 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200087 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174824 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200063 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200053 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200091 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00174816 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20200060 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 18736 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 298643 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3239317 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 10385 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 49312 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P15105234 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3239317 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3239317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200069 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15055929 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200087 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200063 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200077 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200060 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174820 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200088 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15055929 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200063 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174820 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200088 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200069 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200060 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200087 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20200077 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00174819 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15105234 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P481631 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 162656 | AB023181.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 162656 | AF088030.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 162656 | AK001681.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 162656 | BC153874.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 162656 | CR610787.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 162656 | NM_001042486.1 | refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 162656 | NM_014902.3 | refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 162656 | NM_183006.2 | refseq_mrna_human_NM |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15105234 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P481631 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174817 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174802 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15055929 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174797 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174822 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174795 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174799 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174813 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174810 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15055931 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174793 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174806 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174830 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174831 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174826 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00174807 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15105234 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1564697_PM_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008118.hg.1 | ENST00000626198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008118.hg.1 | ENST00000626198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008118.hg.1 | ENST00000626198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008118.hg.1 | NR_024270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008118.hg.1 | NR_024270.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008118.hg.1 | NR_024270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008118.hg.1 | ENST00000626198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008118.hg.1 | ENST00000626198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008118.hg.1 | ENST00000626198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008118.hg.1 | NR_024270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008118.hg.1 | NR_024270.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008118.hg.1 | NR_024270.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100157952.hg.1 | ENST00000626198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100157952.hg.1 | NR_024270.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100157952.hg.1 | ENST00000626198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100157952.hg.1 | NR_024270.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100157952.hg.1 | TC0100008118.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1564697_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564697_PM_a_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564697_a_at4 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021817 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021817 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 178597 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1690382 | NR_024270.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1690382 | BC006119.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1690382 | BC035497.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1690382 | NR_024270.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1690382 | BC006119.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1690382 | BC035497.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1690382 | NR_024270.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1690382 | BC006119.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1690382 | BC035497.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1690382 | NR_024270.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1690382 | BC006119.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1690382 | BC035497.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690382 | NR_024270.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690382 | BC006119.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690382 | BC035497.2 | human_transcriptome |

LINC01144
Genomic Map