Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243211.1 | NP_001230140.1 | 3 - 4 | 431 | 115 | |
| NM_001562.3 | NP_001553.1 | 4 - 5 | 443 | 115 | ||
| XM_011542805.1 | XP_011541107.1 | 3 - 4 | 439 | 115 | ||
| XM_011542806.2 | XP_011541108.1 | 4 - 5 | 447 | 115 | ||
| GenBank mRNA | AF077611.1 | - | 3 - 4 | 223 | 115 | |
| AF380360.1 | - | 4 - 5 | 275 | 115 | ||
| AY044641.1 | - | 4 - 5 | 320 | 115 | ||
| AY266351.1 | - | 3 - 4 | 388 | 115 | ||
| BC007007.1 | - | 4 - 5 | 418 | 3686124 | 115 | |
| BC007461.1 | - | 4 - 5 | 443 | 3681009 | 115 | |
| CR541973.1 | - | 3 - 4 | 223 | 115 | ||
| CR542001.1 | - | 3 - 4 | 223 | 115 | ||
| D49950.1 | - | 4 - 5 | 400 | 115 | ||
| U90434.1 | - | 3 - 4 | 220 | 115 | ||
More Information
Set Membership: |
|

IL18
Genomic Map