Assay Details
| Gene Symbol: TAF5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025247.1 | NP_001020418.1 | 2 - 3 | 306 | 134 | |
| NM_014409.3 | NP_055224.1 | 2 - 3 | 306 | 134 | ||
| XM_005273099.4 | XP_005273156.1 | 2 - 3 | 259 | 134 | ||
| XM_005273100.2 | XP_005273157.1 | 2 - 3 | 197 | 134 | ||
| XM_011544163.1 | XP_011542465.1 | 2 - 3 | 292 | 134 | ||
| XM_017001045.1 | XP_016856534.1 | 2 - 3 | 223 | 134 | ||
| GenBank mRNA | AF069736.1 | - | 1 - 2 | 140 | 134 | |
| BC041094.1 | - | 2 - 3 | 229 | 5551246 | 134 | |
| BE646579.1 | - | 2 - 3 | 297 | 134 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003951.hg
Additional Information:
This assay is in the 5'UTR region of the XM_005273100.2,XM_011544163.1,XM_017001045.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01059900.hg.1 | TC01003951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059900.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059900.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059903.hg.1 | TC01003951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059903.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059903.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059904.hg.1 | TC01003951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059904.hg.1 | ENST00000258281.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059904.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059904.hg.1 | ENST00000366676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059904.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059904.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059901.hg.1 | TC01003951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059901.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059901.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059907.hg.1 | TC01003951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059907.hg.1 | ENST00000258281.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059907.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059907.hg.1 | ENST00000366676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059907.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059907.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059907.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233418.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233418.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233418.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233418.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233418.hg.1 | TC0100017668.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233422.hg.1 | TC0100017668.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100233420.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233420.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233420.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233420.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233420.hg.1 | TC0100017668.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233425.hg.1 | TC0100017668.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000258281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000366676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | ENST00000477957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017668.hg.1 | NM_014409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233419.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233419.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233419.hg.1 | ENST00000366675.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100233419.hg.1 | NM_001025247.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100233419.hg.1 | TC0100017668.hg.1 | Clariom D TC |

TAF5L
Genomic Map