Assay Details
| Gene Symbol: PKHD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177531.4 | NP_803875.2 | 34 - 35 | 4307 | 132 | |
| XM_011517371.2 | XP_011515673.1 | 34 - 35 | 4258 | 132 | ||
| XM_017013969.1 | XP_016869458.1 | 34 - 35 | 4258 | 132 | ||
| XM_017013970.1 | XP_016869459.1 | 33 - 34 | 4144 | 132 | ||
| XM_017013971.1 | XP_016869460.1 | 34 - 35 | 4258 | 132 | ||
| XM_017013972.1 | XP_016869461.1 | 32 - 33 | 3955 | 132 | ||
| XM_017013973.1 | XP_016869462.1 | 10 - 11 | 1314 | 132 | ||
| XM_017013974.1 | XP_016869463.1 | 5 - 6 | 814 | 132 | ||
| GenBank mRNA | AY219181.1 | - | 34 - 35 | 4307 | 132 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000668.hg.1
Set Membership: |
|

PKHD1L1
Genomic Map