Assay Details
| Gene Symbol: PKHD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177531.4 | NP_803875.2 | 71 - 72 | 11649 | 153 | |
| XM_011517371.2 | XP_011515673.1 | 71 - 72 | 11600 | 153 | ||
| XM_017013969.1 | XP_016869458.1 | 71 - 72 | 11600 | 153 | ||
| XM_017013970.1 | XP_016869459.1 | 70 - 71 | 11486 | 153 | ||
| XM_017013971.1 | XP_016869460.1 | 71 - 72 | 11438 | 153 | ||
| XM_017013972.1 | XP_016869461.1 | 69 - 70 | 11297 | 153 | ||
| XM_017013973.1 | XP_016869462.1 | 47 - 48 | 8656 | 153 | ||
| XM_017013974.1 | XP_016869463.1 | 42 - 43 | 8156 | 153 | ||
| GenBank mRNA | AK131338.1 | - | 17 - 18 | 2331 | 153 | |
| AL133640.1 | - | 1 - 2 | 89 | 153 | ||
| AY219181.1 | - | 71 - 72 | 11649 | 153 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000668.hg.1
Set Membership: |
|

PKHD1L1
Genomic Map