Assay Details
| Gene Symbol: CAMKK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032294.2 | NP_115670.1 | 6 - 7 | 791 | 82 | |
| NM_172206.1 | NP_757343.1 | 6 - 7 | 737 | 82 | ||
| XM_011524033.1 | XP_011522335.1 | 5 - 6 | 689 | 82 | ||
| XM_017025219.1 | XP_016880708.1 | 6 - 7 | 900 | 82 | ||
| XM_017025220.1 | XP_016880709.1 | 6 - 7 | 1351 | 82 | ||
| XM_017025222.1 | XP_016880711.1 | 6 - 7 | 968 | 82 | ||
| GenBank mRNA | AF425232.1 | - | 6 - 7 | 744 | 82 | |
| AF425301.1 | - | 6 - 7 | 737 | 82 | ||
| AL136576.1 | - | 6 - 7 | 791 | 82 | ||
| BC043487.1 | - | 6 - 7 | 767 | 5751570 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011524033.1 transcripts.
Set Membership: |
|

CAMKK1
Genomic Map