Assay Details
| Gene Symbol: MAP3K5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005923.3 | NP_005914.1 | 1 - 2 | 813 | 59 | |
| XM_011535839.2 | XP_011534141.2 | 2 - 3 | 1137 | 59 | ||
| XM_017010870.1 | XP_016866359.1 | 2 - 3 | 1136 | 59 | ||
| XM_017010871.1 | XP_016866360.1 | 2 - 3 | 1136 | 59 | ||
| XM_017010872.1 | XP_016866361.1 | 2 - 3 | 1135 | 59 | ||
| XM_017010873.1 | XP_016866362.1 | 2 - 3 | 1134 | 59 | ||
| XM_017010874.1 | XP_016866363.1 | 2 - 3 | 1136 | 59 | ||
| XM_017010875.1 | XP_016866364.1 | 2 - 3 | 1135 | 59 | ||
| GenBank mRNA | AB209093.1 | - | 1 - 2 | 917 | 59 | |
| AK310942.1 | - | 2 - 3 | 592 | 59 | ||
| BC054503.2 | - | 1 - 2 | 760 | 6143181 | 59 | |
| BC088829.1 | - | 1 - 2 | 814 | 6007002 | 59 | |
| D84476.2 | - | 1 - 2 | 719 | 59 | ||
| U67156.1 | - | 1 - 2 | 812 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002146.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
response to ischemia
protein phosphorylation
JNK cascade
activation of JUN kinase activity
intrinsic apoptotic signaling pathway in response to oxidative stress
viral process
response to endoplasmic reticulum stress
p38MAPK cascade
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of JUN kinase activity
innate immune response
positive regulation of myoblast differentiation
positive regulation of JNK cascade
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to hydrogen peroxide
apoptotic signaling pathway
programmed necrotic cell death
positive regulation of neuron death
cellular response to reactive nitrogen species
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
protein kinase binding
protein phosphatase binding
protein homodimerization activity
MAPK cascade
activation of MAPKK activity
response to ischemia
protein phosphorylation
JNK cascade
activation of JUN kinase activity
intrinsic apoptotic signaling pathway in response to oxidative stress
viral process
response to endoplasmic reticulum stress
p38MAPK cascade
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of JUN kinase activity
innate immune response
positive regulation of myoblast differentiation
positive regulation of JNK cascade
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to hydrogen peroxide
apoptotic signaling pathway
programmed necrotic cell death
positive regulation of neuron death
cellular response to reactive nitrogen species
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
protein kinase binding
protein phosphatase binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06027082.hg.1 | TC06002146.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027082.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027082.hg.1 | ENST00000367768.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027082.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027086.hg.1 | TC06002146.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027086.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027086.hg.1 | ENST00000367768.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027086.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027084.hg.1 | TC06002146.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027084.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027084.hg.1 | ENST00000367768.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027084.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027087.hg.1 | TC06002146.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027087.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027087.hg.1 | ENST00000367768.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027087.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000463140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000463140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | ENST00000463140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191970.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191970.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191970.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191970.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191970.hg.1 | TC0600013280.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600191969.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191969.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191969.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191969.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191969.hg.1 | TC0600013280.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600191964.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191964.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191964.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191964.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191964.hg.1 | TC0600013280.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000463140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000463140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | ENST00000463140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013280.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191967.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191967.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191967.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191967.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191967.hg.1 | TC0600013280.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600191968.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191968.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191968.hg.1 | ENST00000359015.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191968.hg.1 | NM_005923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191968.hg.1 | TC0600013280.hg.1 | Clariom D TC |

MAP3K5
Genomic Map