Assay Details
Gene Symbol: SEC11C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033280.3 | NP_150596.1 | 4 - 5 | 555 | 125 | |
XM_011526260.2 | XP_011524562.1 | 5 - 6 | 562 | 125 | ||
XM_017026073.1 | XP_016881562.1 | 4 - 5 | 416 | 125 | ||
GenBank mRNA | AF212233.1 | - | 4 - 5 | 520 | 125 | |
AK314107.1 | - | 4 - 5 | 527 | 125 | ||
BC009703.1 | - | 4 - 5 | 507 | 3895819 | 125 | |
BM760252.1 | - | 2 - 3 | 146 | 125 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000210.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18002432.hg.1 | ENST00000299714.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18002432.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18002432.hg.1 | TC18000210.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18002433.hg.1 | ENST00000299714.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18002433.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18002433.hg.1 | TC18000210.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800151179.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151179.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151179.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151179.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800151179.hg.1 | TC1800007446.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800151179.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151179.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151179.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800151179.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000299714.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000509791.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000585864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000587834.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000588875.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000591406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000592774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | ENST00000593132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_001307941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007446.hg.1 | NM_033280.3 | Refseq |
